KEY311758702 Key Organics / BIONET

methyl 2-methylpyrimidine-5-carboxylate

  • MDL number MFCD11111634
  • Molecular Weight : 152.153

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74 - 76
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106800

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/NE-0701_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart