KEY311758712 Key Organics / BIONET

1-benzyl-4-hydroxypiperidine-4-carboxamide

  • MDL number MFCD17947481
  • Cas Number : 27771-25-1
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 178 - 180
Iupac Name 1-benzyl-4-hydroxy-4-piperidinecarboxamide
Inchi Code 1S/C13H18N2O2/c14-12(16)13(17)6-8-15(9-7-13)10-11-4-2-1-3-5-11/h1-5,17H,6-10H2,(H2,14,16)
Inchi Key YZVZPVSOBJCEJS-UHFFFAOYSA-N

Description

Melting Point 178 - 180
Purity 95%
Storage temp. Store under N2
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94712

Safety Information

signalword Warning
pcodes P422, P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AE-0768_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart