KEY311758719 Key Organics / BIONET

(1S)-1-(2,6-dichloro-3-fluorophenyl)ethan-1-ol

  • MDL number MFCD09863793
  • Cas Number : 877397-65-4
  • Molecular Weight : 209.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108627

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4349_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart