KEY320856983 Key Organics / BIONET

N-benzyl-2-(trifluoromethyl)pyridin-4-amine

  • MDL number MFCD18803520
  • Cas Number : 1280787-28-1
  • Molecular Weight : 252.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104 - 106
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94729

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AF-0227_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart