KEY320857006 Key Organics / BIONET

2-(2-bromoethyl)-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one

  • MDL number MFCD18651760
  • Cas Number : 1272756-54-3
  • Molecular Weight : 257.087

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91380

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8Z-0814_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart