KEY320857013 Key Organics / BIONET

ethyl 2-amino-4-methyl-1,3-oxazole-5-carboxylate

  • MDL number MFCD09028430
  • Cas Number : 79221-15-1
  • Molecular Weight : 170.168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215 - 217
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95178

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BF-0722_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart