KEY320857032 Key Organics / BIONET

ethyl 4-methyl-2-(methylamino)-1,3-thiazole-5-carboxylate hydrochloride

  • MDL number MFCD18380662
  • Cas Number : 1269152-25-1
  • Molecular Weight : 236.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220 - 222
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95201

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BF-0752_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart