KEY320857033 Key Organics / BIONET

methyl 4-amino-3-bromo-5-fluorobenzoate

  • MDL number MFCD11846084
  • Cas Number : 1123171-91-4
  • Molecular Weight : 248.051

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101 - 102
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95202

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BF-0753_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart