KEY320857053 Key Organics / BIONET

ethyl 3-iodo-5-nitro-1H-indole-2-carboxylate

  • MDL number MFCD18651761
  • Cas Number : 494854-46-5
  • Molecular Weight : 360.107

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 276 - 277
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94756

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AF-0734_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart