KEY320857055 Key Organics / BIONET

ethyl (2Z)-2-cyano-3-hydroxy-3-(4-methylphenyl)prop-2-enoate

  • MDL number MFCD18651762
  • Cas Number : 91024-54-3
  • Molecular Weight : 231.251

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43 - 45
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94760

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AF-0742_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart