KEY320857058 Key Organics / BIONET

ethyl 2-(5-chloro-1,3-benzoxazol-2-yl)acetate

  • MDL number MFCD04210543
  • Cas Number : 138420-09-4
  • Molecular Weight : 239.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91 - 92
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95802

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CF-0704_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart