KEY320857060 Key Organics / BIONET

2-(ethoxycarbonyl)-1H-indole-3-carboxylic acid

  • MDL number MFCD18803524
  • Cas Number : 441800-93-7
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 262 - 263
Iupac Name 2-(ethoxycarbonyl)-1H-indole-3-carboxylic acid
Inchi Code 1S/C12H11NO4/c1-2-17-12(16)10-9(11(14)15)7-5-3-4-6-8(7)13-10/h3-6,13H,2H2,1H3,(H,14,15)
Inchi Key HFKAHXLZJKJMKW-UHFFFAOYSA-N

Description

Melting Point 262 - 263
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95804

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/CF-0707_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart