KEY320857075 Key Organics / BIONET

ethyl 6-bromo-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate

  • MDL number MFCD18803526
  • Cas Number : 894851-71-9
  • Molecular Weight : 297.108

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 264 - 266
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95821

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CF-0735_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart