KEY329173246 Key Organics / BIONET

4-[(pyridin-2-yl)methoxy]benzaldehyde

  • MDL number MFCD00469762
  • Cas Number : 57748-41-1
  • Molecular Weight : 213.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 72 - 75
Iupac Name 4-(2-pyridinylmethoxy)benzaldehyde
Inchi Code 1S/C13H11NO2/c15-9-11-4-6-13(7-5-11)16-10-12-3-1-2-8-14-12/h1-9H,10H2
Inchi Key MKHNUKSZFMWQJL-UHFFFAOYSA-N

Description

Melting Point 72 - 75
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68792

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/2P-903_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart