KEY329173247 Key Organics / BIONET

1-[2-(4-fluorophenyl)-4-methyl-1,3-thiazol-5-yl]ethan-1-one

  • MDL number MFCD09295390
  • Cas Number : 923701-97-7
  • Molecular Weight : 235.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106 - 108
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73236

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0845_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart