KEY329173270 Key Organics / BIONET

3-(trifluoromethyl)pyridine-2-carbonitrile

  • MDL number MFCD02091399
  • Cas Number : 406933-21-9
  • Molecular Weight : 172.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 120
Iupac Name 3-(trifluoromethyl)-2-pyridinecarbonitrile
Inchi Code 1S/C7H3F3N2/c8-7(9,10)5-2-1-3-12-6(5)4-11/h1-3H
Inchi Key RDEFTHKSTLWCEU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Store under N2
Physical Form Liquid
Country of Origin GB
Boiling point 120
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96515

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/EF-0235_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart