KEY329173309 Key Organics / BIONET

2-methyl-N-phenylprop-2-enamide

  • MDL number MFCD00048114
  • Cas Number : 1611-83-2
  • Molecular Weight : 161.204

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1863

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10L-706_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart