KEY332645058 Key Organics / BIONET

ethyl 3-azabicyclo[3.1.0]hexane-2-carboxylate

  • MDL number MFCD19686125
  • Cas Number : 75715-99-0
  • Molecular Weight : 155.197

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97045

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FF-0755_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart