KEY332645067 Key Organics / BIONET

7-nitro-1,2-dihydroquinoxalin-2-one

  • MDL number MFCD09999191
  • Cas Number : 89898-96-4
  • Molecular Weight : 191.146

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 281 - 282
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97543

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/GF-0716_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart