KEY332645080 Key Organics / BIONET

tert-butyl N-[(1R,2S)-2-aminocyclohexyl]carbamate >97%ee

  • MDL number MFCD09952106
  • Cas Number : 364385-54-6
  • Molecular Weight : 214.309

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108630

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4352_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart