KEY333430377 Key Organics / BIONET

1-[2-(thiomorpholin-4-yl)ethyl]imidazolidin-2-one

  • MDL number MFCD19982779
  • Cas Number : 1331823-96-1
  • Molecular Weight : 215.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129 - 131
Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97503

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GF-0083_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart