KEY333430385 Key Organics / BIONET

ethyl 2-hydroxy-6-iodo-1,8-naphthyridine-3-carboxylate

  • MDL number MFCD19982781
  • Cas Number : 1330583-61-3
  • Molecular Weight : 344.108

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 289 - 291
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97554

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GF-0733_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart