KEY333430391 Key Organics / BIONET

5-amino-1-tert-butyl-1H-pyrazole-4-carbonitrile

  • MDL number MFCD00128289
  • Cas Number : 158001-28-6
  • Molecular Weight : 164.212

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99 - 101
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97044

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FF-0752_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart