KEY336953416 Key Organics / BIONET

methyl 4-[(morpholin-4-yl)methyl]benzoate

  • MDL number MFCD01167691
  • Cas Number : 68453-56-5
  • Molecular Weight : 235.283

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Barely green clear oil
Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97926

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/HF-0170_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart