KEY336953417 Key Organics / BIONET

methyl 4-[(pyrrolidin-1-yl)methyl]benzoate

  • MDL number MFCD01452504
  • Cas Number : 160598-45-8
  • Molecular Weight : 219.284

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil not distilled
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97927

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/HF-0172_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart