KEY336953419 Key Organics / BIONET

4-[(thiomorpholin-4-yl)methyl]benzoic acid

  • MDL number MFCD00976996
  • Cas Number : 414892-27-6
  • Molecular Weight : 237.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208 - 210
Purity 95
Country of Origin United Kingdom
Product Link http://www.keyorganics.net/bionet/catalog/product/view/id/97929

Safety Information

signalword Warning
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart