KEY336953436 Key Organics / BIONET

4-chloro-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD09837291
  • Cas Number : 104146-17-0
  • Molecular Weight : 147.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 53 - 54
Iupac Name 4-chloro-1,3-thiazole-5-carbaldehyde
Inchi Code 1S/C4H2ClNOS/c5-4-3(1-7)8-2-6-4/h1-2H
Inchi Key GJTWEAFBOIYNLZ-UHFFFAOYSA-N

Description

Melting Point 53 - 54
Purity 95%
Storage temp. Store under N2 at 4
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101107

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/KF-0733_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart