KEY338511735 Key Organics / BIONET

1H-indole-2-carbonitrile

  • MDL number MFCD00187566
  • Cas Number : 36193-65-4
  • Molecular Weight : 142.161

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79929

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5P-050_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart