KEY338511744 Key Organics / BIONET

2-(1-benzylpiperidin-4-ylidene)acetonitrile

  • MDL number MFCD09908274
  • Cas Number : 55022-82-7
  • Molecular Weight : 212.296

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93 - 94
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101091

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KF-0234_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart