KEY338511759 Key Organics / BIONET

7-hydroxy-4-{[4-(hydroxymethyl)piperidin-1-yl]methyl}-2H-chromen-2-one

  • MDL number MFCD20444440
  • Cas Number : 1353878-09-7
  • Molecular Weight : 289.331

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64 - 66
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101899

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LF-0001_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart