KEY338511771 Key Organics / BIONET

1,4-dihydro-1,6-naphthyridin-4-one

  • MDL number MFCD02684596
  • Cas Number : 72754-01-9
  • Molecular Weight : 146.149

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 297 - 299
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102002

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/LF-0708_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart