KEY338511774 Key Organics / BIONET

ethyl 1H-pyrrolo[2,3-b]pyridine-4-carboxylate

  • MDL number MFCD15071133
  • Cas Number : 1261588-72-0
  • Molecular Weight : 190.202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127 - 129
Purity 97
Storage temp. RT
Physical Form White to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102012

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/LF-0736_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart