KEY343107495 Key Organics / BIONET

tert-butyl N-(3-iodo-5-nitrophenyl)carbamate

  • MDL number MFCD20921570
  • Cas Number : 1353878-19-9
  • Molecular Weight : 364.139

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113 - 115
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97020

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/FF-0264_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart