KEY343107500 Key Organics / BIONET

7-chloro-6-hydroxy-5-methyl-2,3-dihydro-1H-isoindol-1-one

  • MDL number MFCD20921592
  • Cas Number : 1353878-10-0
  • Molecular Weight : 197.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239 - 242
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102290

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MF-0019_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart