KEY343107507 Key Organics / BIONET

tert-butyl N-[2-(4-bromophenyl)-2-oxoethyl]carbamate

  • MDL number MFCD20921604
  • Cas Number : 339185-70-5
  • Molecular Weight : 314.179

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96 - 98
Purity 95
Storage temp. Store under N2 at 4°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102326

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P411,P422
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MF-0718_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart