KEY343107511 Key Organics / BIONET

5-(1,3-dioxan-2-yl)-1H-indazol-3-amine

  • MDL number MFCD20921613
  • Cas Number : 218301-24-7
  • Molecular Weight : 219.244

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207 - 209
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106840

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/NF-0716_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart