KEY343107545 Key Organics / BIONET

methyl 4-fluoro-5-methoxy-2-nitrobenzoate

  • MDL number MFCD20921594
  • Cas Number : 159768-50-0
  • Molecular Weight : 229.163

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45 - 47
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102293

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/MF-0023_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart