KEY343107551 Key Organics / BIONET

5-iodo-2-methoxy-4,6-dimethylpyridine-3-carbonitrile

  • MDL number MFCD20526328
  • Cas Number : 858120-02-2
  • Molecular Weight : 288.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 112 - 114
Iupac Name 5-iodo-2-methoxy-4,6-dimethylnicotinonitrile
Inchi Code 1S/C9H9IN2O/c1-5-7(4-11)9(13-3)12-6(2)8(5)10/h1-3H3
Inchi Key KQQZSIHWXLUEMQ-UHFFFAOYSA-N

Description

Melting Point 112 - 114
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94678

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AE-0723_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart