KEY343107572 Key Organics / BIONET

3-bromo-4-chloro-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD08276165
  • Cas Number : 90914-41-3
  • Molecular Weight : 233.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 181 - 183
Iupac Name 3-bromo-4-chloro-1H-pyrazolo[3,4-d]pyrimidine
Inchi Code 1S/C5H2BrClN4/c6-3-2-4(7)8-1-9-5(2)11-10-3/h1H,(H,8,9,10,11)
Inchi Key MDFHFCAMBZNSLV-UHFFFAOYSA-N

Description

Melting Point 181 - 183
Purity 95%
Storage temp. Store at 4
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102333

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411
MSDS https://www.keyorganics.net/media/catalog/product/sds/MF-0733_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart