KEY343107575 Key Organics / BIONET

ethyl 4,5-dichloro-1-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate

  • MDL number MFCD09031524
  • Cas Number : 853105-72-3
  • Molecular Weight : 250.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132 - 134
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102336

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MF-0737_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart