KEY343107576 Key Organics / BIONET

dibromo-1,3,4-thiadiazole

  • MDL number MFCD00466397
  • Cas Number : 55981-29-8
  • Molecular Weight : 243.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 111 - 113
Iupac Name 2,5-dibromo-1,3,4-thiadiazole
Inchi Code 1S/C2Br2N2S/c3-1-5-6-2(4)7-1
Inchi Key MNHUBBRVPVSVCM-UHFFFAOYSA-N

Description

Melting Point 111 - 113
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106817

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/NF-0010_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart