KEY343107577 Key Organics / BIONET

4-ethyl-1-(4-nitrophenyl)-1H-1,2,3-triazole

  • MDL number MFCD20921608
  • Cas Number : 1276538-19-2
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140 - 141
Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106820

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/NF-0019_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart