KEY343107585 Key Organics / BIONET

ethyl 4-chloro-5-nitro-6-oxo-1,6-dihydropyridine-3-carboxylate

  • MDL number MFCD20921575
  • Cas Number : 1210835-74-7
  • Molecular Weight : 246.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 176 - 178
Iupac Name ethyl 4-chloro-5-nitro-6-oxo-1,6-dihydro-3-pyridinecarboxylate
Inchi Code 1S/C8H7ClN2O5/c1-2-16-8(13)4-3-10-7(12)6(5(4)9)11(14)15/h3H,2H2,1H3,(H,10,12)
Inchi Key FETZVQJUBMRFJE-UHFFFAOYSA-N

Description

Melting Point 176 - 178
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97557

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/GF-0737_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart