KEY343107587 Key Organics / BIONET

4-chloro-6-methylpyridine-2-carboxylic acid

  • MDL number MFCD07374951
  • Cas Number : 30235-19-9
  • Molecular Weight : 171.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230 (dec)
Purity 95
Storage temp. RT
Physical Form White to Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106844

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/NF-0723_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart