KEY343107588 Key Organics / BIONET

5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD03422575
  • Cas Number : 1027617-86-2
  • Molecular Weight : 176.142

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75 - 77
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106845

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/NF-0724_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart