KEY343107597 Key Organics / BIONET

(1R,2S,5S)-3-azabicyclo[3.1.0]hexane-2-carboxylic acid >99%ee

  • MDL number MFCD05663832
  • Cas Number : 33294-81-4
  • Molecular Weight : 127.143

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 252 - 256
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108632

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4355_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart