KEY358812820 Key Organics / BIONET

1-(5-bromo-1,3,4-thiadiazol-2-yl)-1,4-diazepane

  • MDL number MFCD21333199
  • Cas Number : 1357147-39-7
  • Molecular Weight : 263.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52 - 54
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94773

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AG-0044_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart