KEY358812821 Key Organics / BIONET

(6-bromo-3-methoxy-2-methylphenyl)methanol

  • MDL number MFCD21333200
  • Cas Number : 110451-91-7
  • Molecular Weight : 231.089

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87 - 90
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94774

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AG-0055_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart