KEY358812832 Key Organics / BIONET

ethyl 2,6-bis(trifluoromethyl)pyridine-4-carboxylate

  • MDL number MFCD17015230
  • Cas Number : 1280520-88-8
  • Molecular Weight : 287.161

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41 - 43
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7398

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11Z-0842_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart