KEY358812833 Key Organics / BIONET

4-[(5-bromo-1,3,4-thiadiazol-2-yl)oxy]benzonitrile

  • MDL number MFCD20368328
  • Cas Number : 1341498-87-0
  • Molecular Weight : 282.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110 - 112
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94775

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AG-0063_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart